Trypan blue 0.4% in aqueous solution ready-to-use, sterile

Supplier: VWR
Warning

K940-100ML
97063-702EA 56.48 USD
97063-702
Trypan blue 0.4% in aqueous solution ready-to-use, sterile
Trypan Blue

A pre-mixed dye solution used in cell culture applications to determine cell viability. A researcher can remove a sample of cells from culture and combine in a 1:1 ratio with the Trypan Blue solution. Under the microscope, dead cells will appear a blue color and viable cells will appear clear and translucent. Using a hemacytometer (gridded microscope slide), a researcher can quantify the percentage of dead cells within a population. Stained cells are ready for counting within five minutes.

Formula: C₃₄H₂₄N₆Na₄O₁₄S₄
MDL Number: MFCD00003969
CAS Number: 72-57-1

Order Now

Specification Test Results

Abs.@ Lambda Max (1:100, Water) 1 - 1.3
Lambda Max (1:100, Water) 585 nm - 595 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR