VWR® 50X TRIS-Acetate-EDTA Buffer, Ultra Pure Grade
Supplier: VWR
K915-4L K915-1.6L K915-5L
97063-696EA 462.36 USD
97063-696 97063-692 97063-698
VWR® 50X TRIS-Acetate-EDTA Buffer, Ultra Pure Grade
Buffers Electrophoresis Buffers Running Buffers
TAE is an extensively used buffer for agarose gel electrophoresis applications requiring high resolution and separation of high molecular weight, double-stranded DNA
TAE is more compatible with in-gel manipulations and band recovery procedures than a TBE buffer. A single strength (1X) solution contains 0.04M Tris-Acetate and 0.001M EDTA with a final pH of 8.0. Low ionic strength. Better resolution of large DNA fragments (>12kbDNA).