VWR® Phosphate Buffered Saline (PBS), 10X, Ultra Pure Grade
Supplier: VWR
K813-500ML
97063-660EA 147.66 USD
97063-660
VWR® Phosphate Buffered Saline (PBS), 10X, Ultra Pure Grade
Buffers Cell Culture Buffers
1X Solution Composition: 137 mM NaCl, 2.7 mM KCl, 9.5 mM Phosphate buffer.
  • Dilute 1:10 in sterile water for making 1X solution
  • 10X solution is supplied 0.2 um filter.
  • pH at 25°C (1X PBS): 6.6 – 7.2
  • DNase, RNase, Protease none detected
  • Bioburden Pass (USP)
  • Suitable for cell culture
  • Does not contain magnesium or calcium