Lithium chloride 8 M in aqueous solution, Biotechnology Grade for biotechnology

Supplier: VWR
Warning

K445-100ML
97063-548EA 172.81 USD
97063-548 97063-546
Lithium chloride 8 M in aqueous solution, Biotechnology Grade for biotechnology
Lithium chloride

A convenient, ready-to-use solution that helps to remove inhibitors of cDNA synthesis and translation.


  • High concentration for easy dilution
  • Nuclease and protease free
  • Does not precipitate DNA, protein, or carbohydrates


This Lithium Chloride Solution comes especially formulated for RT PCR preparation and cDNA synthesis. This solution is a nuclease-free, protease-free, 8 Molar preparation of lithium chloride in deionized water and is highly concentrated for easy dilution.
It is ideal for the precipitation of RNA right after total RNA isolation or in vitro transcription. The lithium chloride is the key to this process, as it efficiently precipitates the RNA, but does not precipitate the DNA, protein, or free nucleotide triphosphates.


To ensure a quality product throughout, this 8M Lithium Chloride Solution is 0.2 micron filtered and packaged in a PETG Nalge bottle. This product is a convenient, ready-to-pipette solution (instead of a crystalline solid) that helps to remove inhibitors of cDNA synthesis and translation.

Formula: LiCl
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3

Order Now

Specification Test Results

DNase none detected
Protease none detected
RNase none detected
Titration 7.7 Molar - 8.2 Molar
Appearance Clear, colorless liquid.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR