Hydroxyethyl cellulose, high purity

Supplier: VWR
Warning

K391-500G
97063-538EA 329.85 USD
97063-538
Hydroxyethyl cellulose, high purity
Hydroxyethyl cellulose
Storage Temperature: Ambient
MDL Number: MFCD00072770
CAS Number: 9004-62-0

Order Now

Specification Test Results

pH (2%, Water) @25°C 6 - 7


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR