L(-)-Cystine USP
Supplier: VWR
J993-100G
97063-470CS
1546.98
USD
97063-470
L(-)-Cystine USP
L(-)-Cystine
White crystals or crystalline powder.
Formula:
HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH Boiling Pt: 428 °C (1013 hPa) Melting Pt: 261…262 °C Density: 1.677 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064228 CAS Number: 56-89-3 EINECS: 200-296-3 Merck Index: 13,02811 |
Specification Test Results
Loss on Drying |
|
Residue on Ignition |
|
Specific Rotation | (-225°) - (-215°) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...