VWR® Yeast Media
Supplier: VWR
J903-500G
97063-450CS 922.92 USD
97063-450 97063-446
VWR® Yeast Media
Media for Microbiology Dehydrated Media for Microbiology
For researchers working with yeast, yeast media (YM and YPD) are available in premixed powder form.
YPD Broth is supplied as a premixed powder, which when dissolved at 50 g per liter of water will yield the formulation-Peptone (20 g/L), Yeast Extract (10 g/L), Glucose (20 g/L) with a final pH of 6.5 ±0.5 at 25°C.Autoclave or sterile filter.
YM is Used for the preparation of aciduric microorganisms like yeasts and molds. Increases transformation efficiency during electroporation of Agrobacterium. Directions for Use: Dissolve 11.2 g premixed powder in 1 L deionized water. Autoclave or sterile filter.Composition per L: 10 g mannitol, 0.5 g potassium phosphate dibasic, 0.4 g yeast extract, 0.2 g magnesium sulfate anhydrous, 0.1 g sodium chloride with a final pH of 7.8-8.2 at 25°C