VWR® TRIS-Borate-EDTA Buffer, 5X Concentrate, Ultra Pure Grade
Supplier: VWR
J885-1L J885-4L J885-5L
97063-432EA 52.36 USD
97063-432 97063-436 97063-438
VWR® TRIS-Borate-EDTA Buffer, 5X Concentrate, Ultra Pure Grade
Buffers Electrophoresis Buffers Running Buffers
TBE is optimized for DNA electrophoresis techniques using both acrylamide and agarose
  • High Buffering Capacity
  • High Ionic Strength
  • Permits Extended Electrophoresis Runs with no Buffer Recirculation
  • Convenient Packaging Options Minimize Weighing and Mixing
After dilution the composition of (1X) Solution contains 0.089 M Tris Base, 0.089 M Borate and 0.002 M EDTA, Disodium Salt, Dihydrate; final pH 8.3.