Agarose, LF™ for pulsed field gel electrophoresis (PFGE)

Supplier: VWR

X174-100G
97063-360CS 3531.31 USD
97063-360 97063-362 97063-364
Agarose, LF™ for pulsed field gel electrophoresis (PFGE)
Agarose

Fine white powder.


  • Optimized for large fragment electrophoresis (PFGE)
  • Ideal for resolving large (>20 kb) DNA fragments
  • Nuclease & protease-free


Agarose LF™ is optimized for intermediate and pulsed-field gel electrophoresis (PFGE) applications. The exceptionally low EEO and high gel strength of Agarose LF™ facilitates faster electrophoresis running times in low concentration gels. This agarose is best suited for resolving large (>20 kb) DNA fragments.

Boiling Pt: 1413 °C
Melting Pt: 801 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

DNase none detected
EEO (Electroendosmosis) = 0.06
Endonuclease/ligase inhibitory factors none
Gel Strength = 2000 g/cm2
Gelling Range 37°C - 41°C
Melting Range 93°C - 96°C
Moisture = 8.5 %
Protease none detected
RNase none detected
Sulfate = 0.06 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR