VWR® SOC Outgrowth Media, Biotechnology Grade
Supplier: VWR
N549-6X5ML
97063-206EA 366.78 USD
97063-206 97063-204
VWR® SOC Outgrowth Media, Biotechnology Grade
Media for Microbiology Prepared Media for Microbiology
SOC Media is a rich media that increases transformation efficiency in competent E. coli. After transformation, cells are incubated in SOC to allow expression of antibiotic resistance genes prior to selection.
  • Save time using premixed and ready-to-use media
  • Increases transformation efficiency
  • Allows expression of antibiotic resistance genes
Formulation: Tryptone/Casein Peptone 2.0%, Yeast Extract 0.5%, Sodium Chloride 8.5 mM, Potassium Chloride 2.5 mM, D-Glucose 20.0 mM, Magnesium Chloride Hexahydrate 10.0 mM, Magnesium Sulfate Heptahydrate 10.0 mM
Biotechnology grade.