IPTG (isopropyl-β-D-thiogalactopyranoside), Ultrapure

Supplier: VWR

Synonyms: IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside

J436-100G
97063-174EA 3906.52 USD
97063-174
IPTG (isopropyl-β-D-thiogalactopyranoside), Ultrapure
IPTG (Isopropyl-β-D-thiogalactopyranoside)
Formula: C₉H₁₈O₅S
MW: 238.31 g/mol
Boiling Pt: 402 °C (1013 hPa)
Melting Pt: 109.5…110.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS Number: 367-93-1
EINECS: 206-703-0

Order Now

Specification Test Results

Abs.@300nm (5%, Water) 0.13
Abs.@400nm (5%, Water) 0.06
Dioxane Test Pass
Melting Point 110°C - 114°C
Moisture (KF) 1 %
pH (5%, Water) @25°C 5.0 - 7.0
Purity 99 %
Solubility (5%, Water) Pass
Specific Rotation (1%, Water) -34.5° - -28.5°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR