VWR® Visiglo™ & Visiglo Plus™ Chemiluminescent Substrate Kits
Supplier: VWR
N219-KIT
97063-148EA 1653.16 USD
97063-148 MPN218-KIT
VWR® Visiglo™ & Visiglo Plus™ Chemiluminescent Substrate Kits
Electrophoresis Reagents Western Blotting Reagents
VisiGlo HRP and VisiGlo Plus HRP are luminol-based chemiluminescent substrates designed for the rapid and sensitive detection of peroxidase-labeled conjugates.
  • Highly sensitive substrates
  • Low background without compromising signal intensity
VisiGlo HRP and VisiGlo Plus HRP can be detected using x-ray film and chemiluminescent imagers in blotting applications or using a luminometer for ELISA. VisiGlo Plus HRP is VWR® most sensitive chemiluminescent substrate and is ideal for detecting proteins that are in low abundance.
VisiGlo AP and VisiGlo Plus AP are the chemiluminescent substrates of choice for easy, reliable detection of alkaline-phosphatase in Western blotting or ELISA. Both products are single component, ready-to-use substrates that do not require special blocking solutions. These substrates glow up to 5 days, allowing for repeated exposure or luminosity reading.
They offer increased sensitivity compared to chromogenic substrates in both blotting and ELISA applications.