Carbenicillin disodium salt, Ultra Pure Grade

Supplier: VWR

J358-1G
97063-144EA 75.28 USD
97063-144
Carbenicillin disodium salt, Ultra Pure Grade
Carbenicillin disodium salt

A penicillin derivative that inhibits cell wall synthesis in gram-negative and gram-positive bacteria.


Recommended working concentration: 0.1–30 µg/ml.

Formula: C₁₇H₁₆N₂O₆S·2Na
MDL Number: MFCD00077683
CAS Number: 4800-94-6

Order Now

Specification Test Results

Potency (on anhydrous substance) Min. 770 µg/mg
Identification (Na) Passes test
pH (25°C; 1 % (0.5 g/50 ml)) 6.5 - 8.0
Moisture (KF) Max. 6.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR