VWR® Hypotonic Lysis Buffer
Supplier: VWR
M334-100ML
97063-130EA 143.58 USD
97063-130
VWR® Hypotonic Lysis Buffer
Buffers Extraction Buffers
Cell lysis in detergent-free Hypotonic Lysis buffer and the subsequent isolation of the crude cytoplasmic fraction can be completed in less than 30 minutes
The protocol is compatible with a variety of downstream applications including Western Blotting and ELISA procedures as well as protein assays, which are sensitive to the presence of detergents in the lysis buffer