VWR® PCR DNA Marker
Supplier: VWR
E854-100RXN
97063-054EA
233.16
USD
97063-054
VWR® PCR DNA Marker
Electrophoresis Reagents
Molecular Weight Markers
8 fragments ranging from 50 to 2000 bp for easy band identification
High-resolution marker optimal for size estimation of PCR fragments. Also available pre-mixed with loading dye or with EZ-Vision® One DNA dye.