Fast green FCF, Certifiable Grade

Supplier: VWR
Warning

E772-25G
97062-982EA 219.84 USD
97062-982
Fast green FCF, Certifiable Grade
Fast Green FCF

Bluish-green dye that is soluble in alcohol. Can be used with safranin as a stain for plant tissue and for demonstrating nuclear histones.

Formula: C₃₇H₃₄N₂Na₂O₁₀S₃
Melting Pt: 290 °C
Storage Temperature: Ambient
MDL Number: MFCD00013053
CAS Number: 2353-45-9
EINECS: 219-091-5
Merck Index: 13,03970

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR