VWR® Destaining Bags
Supplier: VWR
E732-25
97062-968EA 156.75 USD
97062-968
VWR® Destaining Bags
Electrophoresis Reagents Stain Removal Reagents
Bags safely remove ethidium bromide from solution during overnight treatment
Each bag extracts up to 5mg of ethidium bromide, minimizing personnel exposure to toxic material. The special absorbent mixture retains dye molecules in a convenient bag for removal and incineration.