Sodium dodecyl sulfate (SDS) 10% in aqueous solution, Ultra Pure Grade

Supplier: VWR

E719-100ML
97062-964EA 93.38 USD
97062-964
Sodium dodecyl sulfate (SDS) 10% in aqueous solution, Ultra Pure Grade
Sodium dodecyl sulphate (SDS)

Clear colorless liquid

Formula: CH₃(CH₂)₁₁OSO₃Na
Density: 1…1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR