Ribonuclease Inhibitor (RNase Inhibitor)
Supplier: VWR
E633-2KU
97062-922EA 317.41 USD
97062-922
Ribonuclease Inhibitor (RNase Inhibitor)
Proteins and Peptides
RNase Inhibitor is used to inhibit the activity of RNases in reaction mixtures for cDNA synthesis and in vitro transcription, as well as for long term storage of valuable RNA samples
  • Activity: 2kU
A unit of this protein will inhibit 50% of the activity of 5 ng of RNase A.