D-(+)-Sucrose 20% in aqueous solution, Proteomics Grade

Supplier: VWR

Synonyms: alpha-D-Glucopyranoside, β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, beta-D-Fructofuranosyl alpha-D-glucopyranoside, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, D-(+)-Saccharose, Sucrose, Sugar

E543-100ML
97062-874EA 118.9 USD
97062-874
D-(+)-Sucrose 20% in aqueous solution, Proteomics Grade
D(+)-Sucrose

Common reagent in routine nucleic acid procedures and electrophoresis loading dyes.

Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
MDL Number: MFCD00006626
CAS Number: 57-50-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Clarity Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR