VWR® dTTP
Supplier: VWR
E542-40UM
97062-872EA
159.56
USD
97062-872
VWR® dTTP
Nucleic Acid Reagents
Nucleotides
Deoxyribonucleotides are the foundation of DNA and it takes thousands of these nucleotides to make up DNA. DNA consists of a sugar molecule (deoxyribose), a nitrogenous base (purine in adenine and guanine, pyrimidine in thymine or cytosine) and a phosphate molecule. VWR® deoxyribonucleotides are ideal for the amplification of DNA.
- Building blocks for the amplification of DNA
- High purity grade