Magnesium chloride 1 M in aqueous solution, Biotechnology Grade

Supplier: VWR

E525-100ML E525-500ML
97062-848EA 85.01 USD
97062-848 97062-850
Magnesium chloride 1 M in aqueous solution, Biotechnology Grade
Magnesium chloride

A reagent for the preparation of competent cells for transformation.

Formula: MgCl₂
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3

Order Now

Specification Test Results

Conductivity (1:100) @25C 2000 - 2400 umhos
Identification PASS / Fail
Insolubles ≤ 0.005 %
Nuclease NONE / Fail


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR