Dextran sulfate sodium salt, M.W. ~ 500,000 50%, Biotechnology Grade

Supplier: VWR

E516-100ML
97062-828EA 238.11 USD
97062-828
Dextran sulfate sodium salt, M.W. ~ 500,000 50%, Biotechnology Grade
Dextran sulphate sodium salt
Formula: (C₆H₇Na₃O₁₄S₃)n
Storage Temperature: Ambient
MDL Number: MFCD00081551
CAS Number: 9011-18-1

Order Now

Specification Test Results

APHA (1:10) Tan to yellow haze-free liquid
DNase none detected
RNase none detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR