Aprotinin
Supplier: VWR
E429-50MG
97062-754EA 522.66 USD
97062-754 97062-752
Aprotinin
Proteins and Peptides
A competitive and reversible serine protease inhibitor of chymotrypsin, trypsin, kallikrein and plasmin
GHS Symbols: GHS08
Signal Word: Danger
Not an inhibitor of Factor Xa and thrombin.