Ethidium bromide in aqueous solution, Biotechnology Grade

Supplier: VWR

E406-5ML
97062-734EA 68.36 USD
97062-734 97062-736
Ethidium bromide in aqueous solution, Biotechnology Grade
Ethidium bromide

Ethidium Bromide solution is a fluorescent stain for visualizing nucleic acids in solution or in electrophoresis gels. The solution is provided at a concentration of 0.625mg/mL. A final working concentration of 0.5g/mL is easily obtained by adding 1 drop of solution to 50 mL of molten agarose, running buffer or staining buffer. Ethidium Bromide solution in dropper bottles may be stored at room temperature for up to 1 year.


Dropper bottles are shatter-resistant and light-proof. No additional hazardous shipping costs are applied to Ethidium Bromide Dropper Bottles. One drop per 50 mL of water equals 0.5ug/mL Ethidium Bromide.

Formula: C₂₁H₂₀BrN₃
MDL Number: MFCD00011724
CAS Number: 1239-45-8
EINECS: 214-984-6

Order Now

Specification Test Results

Abs.@ Lambda Max 0.68 - 0.74
DNase none detected
Lambda Max (8/100 mL Water) 478 – 482 nm
RNase none detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR