VWR® Biotechnology Grade Phosphate Buffered Saline (PBS) Tablets
Supplier: VWR
E404-200TABS
97062-732CS
1181.88
USD
97062-732
97062-730
VWR® Biotechnology Grade Phosphate Buffered Saline (PBS) Tablets
Buffers
Electrophoresis Buffers
Washing Buffers
Pre-measured tablets for convenient preparation of 1X PBS solution, pH 7.3-7.5.
- May be sterilized by filtration or autoclave, if desired.
- Does not contain calcium or magnesium.
Each tablet, when dissolved in 100 mL of water, prepares 1X PBS solution containing 137 mM sodium chloride, 2.7 mM potassium chloride, and 10 mM phosphate buffer.