VWR® DNA Molecular Weight Marker, High Range
Supplier: VWR
E255-150UG
97062-686EA 967.85 USD
97062-686
VWR® DNA Molecular Weight Marker, High Range
Electrophoresis Reagents Molecular Weight Markers
10 fragments ranging from 1,503 to 48,502 bp for easy band identification
Optimal for evaluation fragments >10 kb.