Calcium chloride, anhydrous ≥96% ACS

Supplier: VWR
Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

1B1110-1KG 1B1110-500G 1B1110-2.5KG
97062-586CS 582 USD
97062-586 97062-590 97062-588
Calcium chloride, anhydrous ≥96% ACS
Calcium chloride
Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 772 °C
Density: 2.174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,01659

Order Now

Specification Test Results

Purity 96%
Titrable Free Base = 0.006 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR