Ammonium dihydrogen phosphate ACS

Supplier: VWR

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

0897-1KG 0897-500G
97062-500CS 379.72 USD
97062-500 97062-502 97062-504
Ammonium dihydrogen phosphate ACS
Ammonium dihydrogen phosphate
Formula: NH₄H₂PO₄
MW: 115.03 g/mol
Melting Pt: 190
Density: 1.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1
EINECS: 231-764-5
Merck Index: 13,00547

Order Now

Specification Test Results

Calcium 0.001%
Chloride = 0.0005%
Heavy Metals = 0.0005%
Insolubles = 0.005%
Iron = 0.001%
Magnesium = 0.0005%
Nitrate = 0.001%
pH (5%, Water) @25°C 3.8 - 4.4
Potassium = 0.005%
Purity = 98%
Sodium = 0.005%
Sulfate = 0.01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR