5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate) ≥99%, Ultrapure

Supplier: VWR
Warning

0885-1G
97062-482EA 368.38 USD
97062-482
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate) ≥99%, Ultrapure
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)

Chromogenic substrate for the detection of alkaline phosphatase. Often used with INT or NBT to amplify the sensitivity of staining.

Formula: C₁₅H₁₅BrClN₂O₄P
Melting Pt: 197 °C
Storage Temperature: Freezer
MDL Number: MFCD00040636
CAS Number: 6578-06-9
EINECS: 229-506-1

Order Now

Specification Test Results

Em (290nm) 5700
Moisture = 2%
O.D.@620nm (0.1%, DMF) = 0.005
Purity (HPLC) = 99%
Solubility (0.10%, DMF) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR