Potassium carbonate, anhydrous ACS
Supplier: VWR
|
Warning
|
Synonyms:
Potash, Carbonic acid potassium salt, Carbonic acid, dipotassium salt
0879-1KG
0879-2.5KG
0879-500G
97062-470CS
321.68
USD
97062-470
97062-472
97062-474
Potassium carbonate, anhydrous ACS
Potassium carbonate
Formula:
K₂CO₃ MW: 138.21 g/mol Melting Pt: 891 Density: 2.42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011382 CAS Number: 584-08-7 EINECS: 209-529-3 Merck Index: 13,07702 |
|
Specification Test Results
Calcium |
|
Chloride |
|
Heavy Metals |
|
Insolubles |
|
Iron |
|
Magnesium |
|
Phosphate |
|
Purity |
|
Silica |
|
Sodium |
|
Sulfur Compounds |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...