Sodium hydrogen carbonate ACS

Supplier: VWR

Synonyms: Sodium bicarbonate

0865-1KG 0865-2.5KG 0865-12KG
97062-460CS 165.97 USD
97062-460 97062-462 10799-426
Sodium hydrogen carbonate ACS
Sodium hydrogen carbonate
Formula: NaHCO₃
MW: 84.01 g/mol
Melting Pt: 270 °C
Density: 2.159 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Ammonium 0.0005%
Calcium = 0.02%
Chloride = 0.003%
Heavy Metals (as Pb) = 0.0005%
Insolubles = 0.015%
Iron = 0.001%
Magnesium = 0.005%
Phosphate = 0.001%
Potassium = 0.005%
Purity 99.7% - 100.3%
Sulfur Compounds = 0.003%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR