Ponceau S

Supplier: VWR
Warning

Synonyms: 3-Hydroxy-4-(2-sulfo-4-[4-sulfophenylazo]phenylazo)-2,7-naphthalenedisulfonic acid sodium salt, 3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, 3-Hydroxy-4-((2-sulfo-4-((4-sulfophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulfonic acid, tetrasodium salt, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt

0860-50G 0860-100G
97062-458EA 225.62 USD
97062-458 97062-456
Ponceau S
Ponceau S

A substitute for acid fuschin in Van Gieson stain. Also used for staining proteins after electrophoresis.

Formula: C₂₂H₁₂N₄Na₄O₁₃S₄
MW: 760.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003892
CAS Number: 6226-79-5
EINECS: 228-319-2

Order Now

Specification Test Results

DNase none detected
Em (520nm, Water) = 27000
Loss on Drying = 6 %
Protease none detected
RNase none detected
Solubility (0.1%, Water) pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR