Sodium dodecyl sulfate (SDS) 20% in aqueous solution, Biotechnology Grade

Supplier: VWR
Warning

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

0837-200ML 0837-500ML
97062-440EA 88.61 USD
97062-440 97062-442
Sodium dodecyl sulfate (SDS) 20% in aqueous solution, Biotechnology Grade
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
Density: 1…1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Conductivity 22800 umhos - 27600 umhos
pH @25°C 5 - 8


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR