Cetrimonium bromide, high purity

Supplier: VWR
Danger

Synonyms: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide

0833-1KG 0833-500G
97062-430CS 825.04 USD
97062-430 97062-432
Cetrimonium bromide, high purity
Cetrimonium bromide

A cationic positively-charged quaternary ammonium compounds. Utilized for applications ranging from solubilizing membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis.

Formula: CH₃(CH₂)₁₅N(Br)(CH₃)₃
Melting Pt: 233…237 °C
Flash Pt: 244 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00011772
CAS Number: 57-09-0
EINECS: 200-311-3
UN: 3077
ADR: 9,III
Merck Index: 13,02034

Order Now

Specification Test Results

Purity = 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR