Xylene cyanole FF, Ultrapure

Supplier: VWR
Warning

0819-20G
97062-406EA 99.62 USD
97062-406
Xylene cyanole FF, Ultrapure
Xylene Cyanole FF
Formula: C₂₅H₂₇N₂NaO₆S₂
Melting Pt: 295 °C
Storage Temperature: Ambient
MDL Number: MFCD00040657
CAS Number: 2650-17-1
EINECS: 220-167-5

Order Now

Specification Test Results

DNase none detected
Em (614nm, Water) = 30000
Lambda Max 612 nm - 616 nm
Loss on Drying = 10 %
RNase none


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR