Bromocresol green ACS

Supplier: VWR

0816-25G
97062-404EA 189.04 USD
97062-404 97062-402
Bromocresol green ACS
Bromocresol green
Formula: C₂₁H₁₄Br₄O₅S
Melting Pt: 217…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00005874
CAS Number: 76-60-8
EINECS: 200-972-8
Merck Index: 13,01370
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12b4d4b3d992f8e86746537bf756af442752c853&vid=553ef1836a5b191a43558f8550e78cf2e424446e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Clarity of Solution Pass
Visual Transition Interval (pH 3.8-5.4) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR