VWR® SSC Buffer
Supplier: VWR
0804-4L
97062-382EA 55.72 USD
97062-382 97062-390
VWR® SSC Buffer
Buffers Electrophoresis Buffers Hybridization Buffers
SSC Buffer is a hybridization buffer for Northern and Southern blots
VWR® offers SSC in various convenient forms: a 20X liquid concentrate, a tablet that prepares 100 ml of a 1X solution, and a Ready-Pack™ that prepares 1 L of 20X concentrate.