TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether), Reagent Grade

Supplier: VWR
Danger

0747-4L 0747-1L
97062-308EA 192.99 USD
97062-308 97062-306
TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether), Reagent Grade
TRITON™ X-114 (Polyethylene glycol tert-octylphenyl ether)

Non-ionic - containing no charge (polyoxyethylenes or alkyl glucosides). Non-ionic detergents are suited for solubilizing embrane proteins without altering biological activity. These non-denaturing, gentler detergents are ideal for studying the conformati

Boiling Pt: ∼150 °C (1013 hPa)
Density: 1.055 g/cm³ (20 °C)
Flash Pt: >93 °C
Storage Temperature: Ambient
MDL Number: MFCD00128254
CAS Number: 9036-19-5
UN: 3082
ADR: 9 III

Order Now

Specification Test Results

Cloud Point (1%, Water) 22 °C - 28 °C
Color (APHA) 100
Index of Refraction 1.48 - 1.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR