VWR® Proteinase K
Supplier: VWR
0706-100MG
97062-238EA 199.7 USD
97062-238 97062-240 97062-242
VWR® Proteinase K
Enzymes
Lyophilised powder
Proteinase K is useful for the inactivation of nucleases during the isolation of DNA and RNA. Active over a broad pH range. No DNase or RNase detected.
A non specific protease with a molecular weight of approximately 18 kDa. Exhibits high stability and activity in the presence of SDS, EDTA, and urea, as well as over a wide pH range. Activity >30 units/mg.