D-(-)-Sorbitol ≥96%, high purity

Supplier: VWR

0691-500G 0691-5KG
97062-202CS 214.2 USD
97062-202 97062-204 97062-206
D-(-)-Sorbitol ≥96%, high purity
D(-)-Sorbitol
Formula: HOH₂C(CH(OH))₄CH₂OH
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 98…100 °C
Density: 1.524 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004708
CAS Number: 50-70-4
EINECS: 200-061-5
Merck Index: 13,08797
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=a084778ac67fbdf7e15478c0db755e78cf3d95c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Arsenic 0.0003 %
Chloride 0.005 %
Heavy Metals 0.001 %
Moisture (KF) = 1 %
Purity (Anhydrous) = 96 %
Residue after Ignition 0.1 %
Sulfate 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR