Nalidixic acid

Supplier: VWR
Danger

0677-50G
97062-176EA 155.46 USD
97062-176 97062-178
Nalidixic acid
Nalidixic acid

Nalidixic acid is a bactericidal agent that inhibits DNA synthesis by blocking the activity of DNA gyrase in both gram-positive and gram-negative bacteria Recommended working concentration: 15 µg/ml

Formula: C₁₂H₁₂N₂O₃
MW: 232.24 g/mol
Melting Pt: 227 °C
Storage Temperature: Ambient
MDL Number: MFCD00006884
CAS Number: 389-08-2
EINECS: 206-864-7

Order Now

Specification Test Results

Heavy Metals (as Pb) = 0.002 %
Loss on Drying = 0.5 %
Melting Point 225 °C - 231 °C
Purity = 98 %
Residue on Ignition = 0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR