D-(-)-Ribose ≥98.0%, high purity

Supplier: VWR

0671-100G
97062-164CS 729.48 USD
97062-164 97062-162
D-(-)-Ribose ≥98.0%, high purity
D(-)-Ribose
Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 90…95 °C
Density: 0.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00135453
CAS Number: 50-69-1
EINECS: 200-059-4
Merck Index: 12,08371

Order Now

Specification Test Results

Arsenic 0.0002 %
Ash = 0.2 %
Heavy Metals (as Pb) 0.001 %
Loss on Drying = 3.0 %
Purity 98.0 %
Specific Rotation (4%, Water) (-21.5°) to (-19.7°)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR