Linoleic acid, Ultrapure

Supplier: VWR

Synonyms: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid

0660-10G
97062-128EA 224.14 USD
97062-128
Linoleic acid, Ultrapure
Linoleic acid
Formula: C₁₈H₃₂O₂
MW: 280.45 g/mol
Boiling Pt: 360
Melting Pt: -5
MDL Number: MFCD00064241
CAS Number: 60-33-3
Merck Index: 13,05527

Order Now

Specification Test Results

Purity = 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR