D-(+)-Glucose monohydrate

Supplier: VWR

Synonyms: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate

0643-1KG
97062-088CS 340.29 USD
97062-088 97062-090
D-(+)-Glucose monohydrate
D(+)-Glucose monohydrate
Formula: C₆H₁₂O₆·H₂O
MW: 198.17 g/mol
MDL Number: MFCD00149450
CAS Number: 14431-43-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9ea2af2ca62ca94d86809070593a60aafd8cd1b&vid=e549b499dca7fb95811640efad1429268c8f6d8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

DNase none detected
Insolubles = 0.005 %
Loss on Drying 8 % - 9 %
Purity (Anhydrous) = 99.5 %
RNase none detected
Specific Rotation 52.6 °- 53.2°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR