Ammonium chloride ACS
Supplier: VWR
|
Warning
|
Synonyms:
Ammonium muriate, Salmiac
0621-1KG
0621-2.5KG
0621-500G
97062-046CS
327.94
USD
97062-046
97062-048
97062-050
Ammonium chloride ACS
Ammonium chloride
Formula:
NH₄Cl MW: 53.49 g/mol Melting Pt: 335 °C Density: 1.5256 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011420 CAS Number: 12125-02-9 EINECS: 235-186-4 Merck Index: 13,00510 |
Specification Test Results
Calcium |
|
Heavy Metals (as Pb) |
|
Insolubles |
|
Iron |
|
Magnesium |
|
pH (5%, Water) @25°C | 4.5 - 5.5 |
Phosphate |
|
Purity |
|
Residue on Ignition |
|
Sulfate |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...