Coomassie brilliant blue G-250, Ultrapure

Supplier: VWR

0615-10G 0615-50G 0615-25G
97062-030EA 61.78 USD
97062-030 97062-034 97062-032
Coomassie brilliant blue G-250, Ultrapure
Coomassie brilliant blue G-250

Commonly used stain for the detection of protein bands following electrophoresis.

Formula: C₄₇H₄₈N₃NaO₇S₂
Density: >1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078482
CAS Number: 6104-58-1
EINECS: 228-058-4

Order Now

Specification Test Results

Em (595nm, Water) = 36300
Protein Staining Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR