Formamide, Ultrapure

Supplier: VWR
Danger

Synonyms: Carbamaldehyde

0606-100ML 0606-500ML 0606-950ML
97062-006EA 46.96 USD
97062-006 97062-008 97062-010
Formamide, Ultrapure
Formamide
Formula: HCONH₂
MW: 45.04 g/mol
Boiling Pt: 210
Melting Pt: 2
Density: 1.1334 g/cm³ (20 °C)
Flash Pt: 79
Storage Temperature: Refrigerator
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@280nm = 0.05
Conductivity = 100 umhos
Copper 0.00001 %
DNase none detected
Freezing Range 1 °C - 3 °C
Iron 0.0005 %
Lead 0.00005 %
Moisture (KF) 0.75 %
Purity 99.5 %
RNase none
Zinc 0.00005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR