Safranine O, Certifiable Grade

Supplier: VWR

0574-100G 0574-25G
97061-952EA 341.36 USD
97061-952 97061-954
Safranine O, Certifiable Grade
Safranine O

Part Fleming Triple stain, a chromosome stain prepared with light green SF and in Gram stain.

Formula: C₂₀H₁₉ClN₄
Density: 1.447 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011759
CAS Number: 477-73-6
EINECS: 207-518-8

Order Now

Specification Test Results

Absorbance maximum 527 - 533 nm
Dye Content (Anhydrous) =80%
Loss on Drying = 4%
P-15/P+15 1.1 - 1.32


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR