Bromocresol green sodium salt ACS

Supplier: VWR

0548-50G
97061-890CS 2816.99 USD
97061-890
Bromocresol green sodium salt ACS
Bromocresol green sodium salt

A pH indicator (Yellow to Blue; range 3.8 - 5.4).

Formula: C₂₁H₁₃Br₄NaO₅S
Melting Pt: 225 °C
Storage Temperature: Ambient
MDL Number: MFCD00148898
CAS Number: 62625-32-5
EINECS: 263-657-4

Order Now

Specification Test Results

Insolubles Test None
Visual Transition Interval (pH 3.8-5.4) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR